Tisk…

5,8-Dimethyl-4-hydroxyquinoline

Dodavatel: Apollo Scientific

OR307844-1G OR307844-5G
APOSOR307844-1GEA 6220 CZK
APOSOR307844-1G APOSOR307844-5G
5,8-Dimethyl-4-hydroxyquinoline
5,8-Dimethyl-4-hydroxyquinoline
Vzorec: C₁₁H₁₁NO
MW: 173,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272456
CAS číslo: 203626-57-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR