2-Chloro-7-methylquinoline-3-carbonitrile
Dodavatel: Apollo Scientific
OR307832-1G
OR307832-5G
APOSOR307832-1GEA
8340
CZK
APOSOR307832-1G
APOSOR307832-5G
2-Chloro-7-methylquinoline-3-carbonitrile
2-Chloro-7-methylquinoline-3-carbonitrile
Vzorec:
C₁₁H₇ClN₂ MW: 202,64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08437564 CAS číslo: 79249-34-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...