Tisk…

6-Chloro-4-hydrazinoquinoline hydrochloride

Dodavatel: Apollo Scientific

OR307825-1G OR307825-5G
APOSOR307825-1GEA 8570 CZK
APOSOR307825-1G APOSOR307825-5G
6-Chloro-4-hydrazinoquinoline hydrochloride
6-Chloro-4-hydrazinoquinoline hydrochloride
Vzorec: C₉H₈ClN₃·ClH
MW: 230,094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684136
CAS číslo: 1171478-51-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR