6-Bromo-3H-thieno[2,3-d]pyrimidin-4-one 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Bromothieno[2,3-d]pyrimidin-4(3H)-one
OR307766-5G
APOSOR307766-5GEA
4730
CZK
APOSOR307766-5G
6-Bromo-3H-thieno[2,3-d]pyrimidin-4-one 95%
6-Bromo-3H-thieno[2,3-d]pyrimidin-4-one
Vzorec:
C₆H₃BrN₂OS MW: 231,07 g/mol Bod tání: 299…300 °C Storage Temperature: Ambient |
MDL Number:
MFCD12196499 CAS číslo: 56844-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...