Tisk…

6-Bromo-benzo[d]isoxazole-3-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Bromobenzo[d]isoxazole-3-carboxylic acid

OR307765-1G OR307765-5G OR307765-250MG
APOSOR307765-1GEA 6520 CZK
APOSOR307765-1G APOSOR307765-5G APOSOR307765-250MG
6-Bromo-benzo[d]isoxazole-3-carboxylic acid 95%
6-Bromo-benzo[d]isoxazole-3-carboxylic acid
Vzorec: C₈H₄BrNO₃
MW: 242,026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027541
CAS číslo: 1123169-17-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR