Tisk…

(1-(4-Bromophenyl)cyclopropyl)methanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR307753-1G OR307753-250MG
APOSOR307753-1GEA 11180 CZK
APOSOR307753-1G APOSOR307753-250MG
(1-(4-Bromophenyl)cyclopropyl)methanamine hydrochloride
(1-(4-Bromophenyl)cyclopropyl)methanamine hydrochloride
Vzorec: C₁₀H₁₂BrN·ClH
MW: 262,574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13619674
CAS číslo: 1208915-57-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR