Tisk…

cis-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate

Dodavatel: Apollo Scientific

OR307706-1G OR307706-250MG
APOSOR307706-1GEA 14490 CZK
APOSOR307706-1G APOSOR307706-250MG
cis-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate
cis-tert-Butyl 3-(3-bromophenyl)-aziridine-2-carboxylate
Vzorec: C₁₃H₁₆BrNO₂
MW: 298,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26936317
CAS číslo: 1980008-31-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR