cis-tert-Butyl 3-(4-nitrophenyl)aziridine-2-carboxylate
Dodavatel: Apollo Scientific
OR307704-1G
OR307704-250MG
APOSOR307704-1GEA
14490
CZK
APOSOR307704-1G
APOSOR307704-250MG
cis-tert-Butyl 3-(4-nitrophenyl)aziridine-2-carboxylate
cis-tert-Butyl 3-(4-nitrophenyl)aziridine-2-carboxylate
Vzorec:
C₁₃H₁₆N₂O₄ MW: 264,277 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24368289 CAS číslo: 1431364-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...