Tisk…

cis-tert-Butyl 3-(4-methoxyphenyl)aziridine-2-carboxylate

Dodavatel: Apollo Scientific

OR307703-1G OR307703-250MG
APOSOR307703-1GEA 14490 CZK
APOSOR307703-1G APOSOR307703-250MG
cis-tert-Butyl 3-(4-methoxyphenyl)aziridine-2-carboxylate
cis-tert-Butyl 3-(4-methoxyphenyl)aziridine-2-carboxylate
Vzorec: C₁₄H₁₉NO₃
MW: 249,306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368287
CAS číslo: 1431364-00-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR