Tisk…

trans-tert-Butyl 3-(4-methoxyphenyl)aziridine-2-carboxylate

Dodavatel: Apollo Scientific

OR307702-1G OR307702-250MG
APOSOR307702-1GEA 14490 CZK
APOSOR307702-1G APOSOR307702-250MG
trans-tert-Butyl 3-(4-methoxyphenyl)aziridine-2-carboxylate
trans-tert-Butyl 3-(4-methoxyphenyl)aziridine-2-carboxylate
Vzorec: C₁₄H₁₉NO₃
MW: 249,306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368286
CAS číslo: 1416307-72-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR