Tisk…

trans-tert-Butyl 3-phenylaziridine-2-carboxylate

Dodavatel: Apollo Scientific
Varování

OR307696-1G OR307696-250MG
APOSOR307696-1GEA 14490 CZK
APOSOR307696-1G APOSOR307696-250MG
trans-tert-Butyl 3-phenylaziridine-2-carboxylate
trans-tert-Butyl 3-phenylaziridine-2-carboxylate
Vzorec: C₁₃H₁₇NO₂
MW: 219,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368280
CAS číslo: 925253-03-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR