trans-4-Aminotetrahydrofuran-3-ol hydrochloride
Dodavatel: Apollo Scientific
OR307685-1G
OR307685-250MG
APOSOR307685-1GEA
9920
CZK
APOSOR307685-1G
APOSOR307685-250MG
trans-4-Aminotetrahydrofuran-3-ol hydrochloride
trans-4-Aminotetrahydrofuran-3-ol hydrochloride
Vzorec:
C₄H₉NO₂·ClH Storage Temperature: Ambient |
MDL Number:
MFCD11520524 CAS číslo: 215940-96-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...