(1R,2R,3S,4S)-3-(tert-Butoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Dodavatel: Apollo Scientific
OR307684-1G
OR307684-250MG
APOSOR307684-1GEA
34720
CZK
APOSOR307684-1G
APOSOR307684-250MG
(1R,2R,3S,4S)-3-(tert-Butoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
(1R,2R,3S,4S)-3-(tert-Butoxycarbonylamino)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Vzorec:
C₁₃H₁₉NO₄ MW: 253,294 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24368276 CAS číslo: 871357-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...