Tisk…

cis-(6-Hydroxymethyl-cyclohex-3-enyl)-carbamic acid tert-butyl ester

Dodavatel: Apollo Scientific

OR307679-1G OR307679-250MG
APOSOR307679-1GEA 15150 CZK
APOSOR307679-1G APOSOR307679-250MG
cis-(6-Hydroxymethyl-cyclohex-3-enyl)-carbamic acid tert-butyl ester
cis-(6-Hydroxymethyl-cyclohex-3-enyl)-carbamic acid tert-butyl ester
Vzorec: C₁₂H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD04974360
CAS číslo: 539823-26-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR