(1R,2S)-2-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR307676-1G
OR307676-250MG
APOSOR307676-1GEA
11840
CZK
APOSOR307676-1G
APOSOR307676-250MG
(1R,2S)-2-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 96%
(1R,2S)-2-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid
Vzorec:
C₁₁H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD09750530 CAS číslo: 130981-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...