(1R,2S)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
Dodavatel: Apollo Scientific
OR307647-1G
OR307647-250MG
APOSOR307647-1GEA
33730
CZK
APOSOR307647-1G
APOSOR307647-250MG
(1R,2S)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
(1R,2S)-2-Amino-cyclohex-3-enecarboxylic acid hydrochloride
Vzorec:
C₇H₁₁NO₂·ClH MW: 177,629 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04112725 CAS číslo: 131783-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...