Tisk…

[2,2-Dimethyl-1-(2-oxo-ethyl)-propyl]-carbamic acid tert-butyl ester

Dodavatel: Apollo Scientific

OR307644-1G OR307644-250MG
APOSOR307644-1GEA 40670 CZK
APOSOR307644-1G APOSOR307644-250MG
[2,2-Dimethyl-1-(2-oxo-ethyl)-propyl]-carbamic acid tert-butyl ester
[2,2-Dimethyl-1-(2-oxo-ethyl)-propyl]-carbamic acid tert-butyl ester
Vzorec: C₁₂H₂₃NO₃
MW: 229,316 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437531
CAS číslo: 892874-26-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR