1-(1-Aminobutyl)-naphthalen-2-ol hydrochloride
Dodavatel: Apollo Scientific
OR307642-1G
OR307642-250MG
APOSOR307642-1GEA
15640
CZK
APOSOR307642-1G
APOSOR307642-250MG
1-(1-Aminobutyl)-naphthalen-2-ol hydrochloride
1-(1-Aminobutyl)-naphthalen-2-ol hydrochloride
Vzorec:
C₁₄H₁₇NO·ClH MW: 251,752 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08437529 CAS číslo: 915781-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...