Tisk…

[1-(2-Hydroxy-ethyl)-2,2-dimethyl-propyl]-carbamic acid tert-butyl ester

Dodavatel: Apollo Scientific
Varování

OR307640-1G OR307640-250MG
APOSOR307640-1GEA 18950 CZK
APOSOR307640-1G APOSOR307640-250MG
[1-(2-Hydroxy-ethyl)-2,2-dimethyl-propyl]-carbamic acid tert-butyl ester
[1-(2-Hydroxy-ethyl)-2,2-dimethyl-propyl]-carbamic acid tert-butyl ester
Vzorec: C₁₂H₂₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD08437526
CAS číslo: 892874-24-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR