Tisk…

3-Amino-4-methyl-pentan-1-ol

Dodavatel: Apollo Scientific
Varování

OR307620-1G OR307620-250MG OR307620-5G
APOSOR307620-1GEA 10320 CZK
APOSOR307620-1G APOSOR307620-250MG APOSOR307620-5G
3-Amino-4-methyl-pentan-1-ol
3-Amino-4-methyl-pentan-1-ol
Vzorec: C₆H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD07366611
CAS číslo: 4379-15-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR