1-(Amino-p-tolyl-methyl)-naphthalen-2-ol
Dodavatel: Apollo Scientific
OR307614-1G
OR307614-250MG
APOSOR307614-1GEA
18680
CZK
APOSOR307614-1G
APOSOR307614-250MG
1-(Amino-p-tolyl-methyl)-naphthalen-2-ol
1-(Amino-p-tolyl-methyl)-naphthalen-2-ol
Vzorec:
C₁₈H₁₇NO MW: 263,334 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09260390 CAS číslo: 561052-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...