Tisk…

1-[Amino-(4-bromo-phenyl)-methyl]-naphthalen-2-ol hydrochloride

Dodavatel: Apollo Scientific

OR307611-1G OR307611-250MG
APOSOR307611-1GEA 21060 CZK
APOSOR307611-1G APOSOR307611-250MG
1-[Amino-(4-bromo-phenyl)-methyl]-naphthalen-2-ol hydrochloride
1-[Amino-(4-bromo-phenyl)-methyl]-naphthalen-2-ol hydrochloride
Vzorec: C₁₇H₁₄BrNO·ClH
MW: 364,664 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366594
CAS číslo: 1170560-78-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR