diendo-(3-Amino-bicyclo[2.2.1]hept-2-yl)-methanol hydrochloride
Dodavatel: Apollo Scientific
OR307593-1G
OR307593-250MG
APOSOR307593-1GEA
21990
CZK
APOSOR307593-1G
APOSOR307593-250MG
diendo-(3-Amino-bicyclo[2.2.1]hept-2-yl)-methanol hydrochloride
diendo-(3-Amino-bicyclo[2.2.1]hept-2-yl)-methanol hydrochloride
Vzorec:
C₈H₁₅NO·ClH MW: 177,672 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00143122 CAS číslo: 205639-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...