Tisk…
cis-(6-Amino-cyclohex-3-enyl)-methanol hydrochloride
Dodavatel: Apollo Scientific
OR307590-1G OR307590-250MG
APOSOR307590-1GEA 21990 CZK
APOSOR307590-1G APOSOR307590-250MG
cis-(6-Amino-cyclohex-3-enyl)-methanol hydrochloride
cis-(6-Amino-cyclohex-3-enyl)-methanol hydrochloride
Vzorec: C₇H₁₃NO·ClH
MW: 163,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143989
CAS číslo: 1212171-08-8