Tisk…

(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-acetonitrile hydrochloride

Dodavatel: Apollo Scientific

OR307543-1G OR307543-250MG
APOSOR307543-1GEA 14680 CZK
APOSOR307543-1G APOSOR307543-250MG
(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-acetonitrile hydrochloride
(1,2,3,4-Tetrahydro-isoquinolin-1-yl)-acetonitrile hydrochloride
Vzorec: C₁₁H₁₂N₂·ClH
MW: 208,687 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01529685
CAS číslo: 627098-24-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR