Tisk…

2-Amino-6-chloro-3-methylquinoline

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-6-chloro-3-methyl-1-azanaphthalene, 6-Chloro-3-methylquinolin-2-amine

OR307519-1G OR307519-5G
APOSOR307519-1GEA 9530 CZK
APOSOR307519-1G APOSOR307519-5G
2-Amino-6-chloro-3-methylquinoline
2-Amino-6-chloro-3-methylquinoline
Vzorec: C₁₀H₉ClN₂
MW: 192,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272509
CAS číslo: 137110-42-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR