3-Bromo-5-iodopyrazole 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR307307-1G
OR307307-250MG
OR307307-5G
APOSOR307307-1GEA
6850
CZK
APOSOR307307-1G
APOSOR307307-250MG
APOSOR307307-5G
3-Bromo-5-iodopyrazole 98%
3-Bromo-5-iodopyrazole
Vzorec:
C₃H₂BrIN₂ MW: 272,87 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32203992 CAS číslo: 1015779-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...