Tisk…

Ethyl 3-bromo-1-(1-ethoxyethyl)-1H-pyrazole-4-carboxylate

Dodavatel: Apollo Scientific
Varování

OR307298-1G OR307298-250MG
APOSOR307298-1GEA 2070 CZK
APOSOR307298-1G APOSOR307298-250MG
Ethyl 3-bromo-1-(1-ethoxyethyl)-1H-pyrazole-4-carboxylate
Ethyl 3-bromo-1-(1-ethoxyethyl)-1H-pyrazole-4-carboxylate
Vzorec: C₁₀H₁₅BrN₂O₃
MW: 291,142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30721067
CAS číslo: 1639858-78-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR