4,5-Diiodo-3-methyl-1H-pyrazole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,4-Diiodo-5-methyl-1H-pyrazole
OR307291-1G
OR307291-5G
APOSOR307291-1GEA
992
CZK
APOSOR307291-1G
APOSOR307291-5G
4,5-Diiodo-3-methyl-1H-pyrazole
4,5-Diiodo-3-methyl-1H-pyrazole
Vzorec:
C₄H₄I₂N₂ MW: 333,897 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20484310 CAS číslo: 6715-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...