3-Bromo-1-(1-ethoxyethyl)-4-nitro-1H-pyrazole
Dodavatel: Apollo Scientific
|
Varování
|
OR307290-1G
OR307290-250MG
APOSOR307290-1GEA
2070
CZK
APOSOR307290-1G
APOSOR307290-250MG
3-Bromo-1-(1-ethoxyethyl)-4-nitro-1H-pyrazole
3-Bromo-1-(1-ethoxyethyl)-4-nitro-1H-pyrazole
Vzorec:
C₇H₁₀BrN₃O₃ MW: 264,077 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30721068 CAS číslo: 1639858-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...