3-Iodo-4-chloro-1H-pyrazole
Dodavatel: Apollo Scientific
|
Varování
|
OR307259-1G
OR307259-250MG
APOSOR307259-1GEA
8070
CZK
APOSOR307259-1G
APOSOR307259-250MG
3-Iodo-4-chloro-1H-pyrazole
3-Iodo-4-chloro-1H-pyrazole
Vzorec:
C₃H₂ClIN₂ MW: 228,419 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28975224 CAS číslo: 27258-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...