Tisk…

1-Chloro-4-cyclopropylbenzene

Dodavatel: Apollo Scientific

OR307247-1G OR307247-5G
APOSOR307247-1GEA 11180 CZK
APOSOR307247-1G APOSOR307247-5G
1-Chloro-4-cyclopropylbenzene
1-Chloro-4-cyclopropylbenzene
Vzorec: C₉H₉Cl
MW: 152,621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527177
CAS číslo: 1798-84-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR