2-(Furan-3-yl)acetic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR307188-1G
OR307188-250MG
OR307188-100MG
APOSOR307188-1GEA
9860
CZK
APOSOR307188-1G
APOSOR307188-250MG
APOSOR307188-100MG
2-(Furan-3-yl)acetic acid
2-(Furan-3-yl)acetic acid
Vzorec:
C₆H₆O₃ MW: 126,11 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01859888 CAS číslo: 123617-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...