Tisk…

tert-Butyl [(3S)-5-oxotetrahydrofuran-3-yl]carbamate

Dodavatel: Apollo Scientific
Varování

OR307116-5G OR307116-25G
APOSOR307116-5GEA 3640 CZK
APOSOR307116-5G APOSOR307116-25G
tert-Butyl [(3S)-5-oxotetrahydrofuran-3-yl]carbamate
tert-Butyl [(3S)-5-oxotetrahydrofuran-3-yl]carbamate
Vzorec: C₉H₁₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD04113636
CAS číslo: 104227-71-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR