Tisk…

(2R)-Piperazine-2-carboxylic acid, N4-BOC protected 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (R)-1-Boc-Piperazine-3-carboxylic acid

OR307035-5G OR307035-1G OR307035-25G
APOSOR307035-5GEA 5060 CZK
APOSOR307035-5G APOSOR307035-1G APOSOR307035-25G
(2R)-Piperazine-2-carboxylic acid, N4-BOC protected 97%
(2R)-Piperazine-2-carboxylic acid, N4-BOC protected
Vzorec: C₁₀H₁₈N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD02179099
CAS číslo: 192330-11-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR