3-Bromo-5-azaindole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromo-1H-pyrrolo[3,2-c]pyridine
OR30681-1G
OR30681-250MG
OR30681-5G
APOSOR30681-1GEA
2690
CZK
APOSOR30681-1G
APOSOR30681-250MG
APOSOR30681-5G
3-Bromo-5-azaindole
3-Bromo-5-azaindole
Vzorec:
C₇H₅BrN₂ MW: 197,032 g/mol Bod tání: 184…190 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08690126 CAS číslo: 23612-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...