6-Cyclopropylnicotinic acid 95+%
Dodavatel: Apollo Scientific
OR306620-1G
OR306620-100MG
OR306620-250MG
APOSOR306620-1GEA
11010
CZK
APOSOR306620-1G
APOSOR306620-100MG
APOSOR306620-250MG
6-Cyclopropylnicotinic acid 95+%
6-Cyclopropylnicotinic acid
Vzorec:
C₉H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18255573 CAS číslo: 75893-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...