Tisk…

3-Methyl-1H-indole-4-carboxylic acid 96+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Carboxy-3-methyl-1H-indole

OR30661-100MG OR30661-1G OR30661-250MG
APOSOR30661-100MGEA 1450 CZK
APOSOR30661-100MG APOSOR30661-1G APOSOR30661-250MG
3-Methyl-1H-indole-4-carboxylic acid 96+%
3-Methyl-1H-indole-4-carboxylic acid
Vzorec: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11869751
CAS číslo: 1159511-18-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR