Tisk…

2-Cyclopropylpyridin-4-amine 95+%

Dodavatel: Apollo Scientific
Nebezpečí

OR306608-1G OR306608-250MG
APOSOR306608-1GEA 12470 CZK
APOSOR306608-1G APOSOR306608-250MG
2-Cyclopropylpyridin-4-amine 95+%
2-Cyclopropylpyridin-4-amine
Vzorec: C₈H₁₀N₂
MW: 134,178 g/mol
Bod tání: 74…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12032771
CAS číslo: 340006-71-5
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR