Tisk…

6-Cyclopropylpyridin-3-amine

Dodavatel: Apollo Scientific
Nebezpečí

OR306607-1G OR306607-250MG OR306607-5G
APOSOR306607-1GEA 5720 CZK
APOSOR306607-1G APOSOR306607-250MG APOSOR306607-5G
6-Cyclopropylpyridin-3-amine
6-Cyclopropylpyridin-3-amine
Vzorec: C₈H₁₀N₂
MW: 134,178 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12033370
CAS číslo: 1159821-66-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR