Tisk…

4-Cyclopropylpyridin-3-amine

Dodavatel: Apollo Scientific
Varování

OR306606-1G OR306606-250MG OR306606-5G
APOSOR306606-1GEA 16370 CZK
APOSOR306606-1G APOSOR306606-250MG APOSOR306606-5G
4-Cyclopropylpyridin-3-amine
4-Cyclopropylpyridin-3-amine
Vzorec: C₈H₁₀N₂
Storage Temperature: Refrigerator
MDL Number: MFCD22379683
CAS číslo: 1365763-16-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR