2-Cyclopropylpyridin-3-amine
Dodavatel: Apollo Scientific
|
Varování
|
OR306605-1G
OR306605-250MG
OR306605-5G
APOSOR306605-1GEA
10320
CZK
APOSOR306605-1G
APOSOR306605-250MG
APOSOR306605-5G
2-Cyclopropylpyridin-3-amine
2-Cyclopropylpyridin-3-amine
Vzorec:
C₈H₁₀N₂ MW: 134,178 g/mol Bod tání: 173 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD23143908 CAS číslo: 1417519-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...