Tisk…

3-(Hydroxymethyl)pyrrolidin-3-ol hydrochloride 96%

Dodavatel: Apollo Scientific

OR306500-1G OR306500-500MG
APOSOR306500-1GEA 24800 CZK
APOSOR306500-1G APOSOR306500-500MG
3-(Hydroxymethyl)pyrrolidin-3-ol hydrochloride 96%
3-(Hydroxymethyl)pyrrolidin-3-ol hydrochloride
Vzorec: C₅H₁₁NO₂·ClH
MW: 153,607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26096974
CAS číslo: 1439900-52-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR