Tisk…

2-(Aminomethyl)-1,3-thiazole-4-carboxylic acid, 2-BOC protected

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-{[(tert-Butoxycarbonyl)amino]methyl}-1,3-thiazole-4-carboxylic acid

OR30643-1G OR30643-250MG
APOSOR30643-1GEA 13990 CZK
APOSOR30643-1G APOSOR30643-250MG
2-(Aminomethyl)-1,3-thiazole-4-carboxylic acid, 2-BOC protected
2-(Aminomethyl)-1,3-thiazole-4-carboxylic acid, 2-BOC protected
Vzorec: C₁₀H₁₄N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD10703327
CAS číslo: 71904-80-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR