Tisk…

3-Methyloxetan-3-amine hydrochloride 97%

Dodavatel: Apollo Scientific

OR306108-1G OR306108-25G OR306108-5G
APOSOR306108-1GEA 4570 CZK
APOSOR306108-1G APOSOR306108-25G APOSOR306108-5G
3-Methyloxetan-3-amine hydrochloride 97%
3-Methyloxetan-3-amine hydrochloride
Vzorec: C₄H₉NO·ClH
MW: 123,581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17676116
CAS číslo: 1363404-87-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR