Tisk…

6-(Bromomethyl)isoquinoline hydrobromide

Dodavatel: Apollo Scientific
Varování

OR30570-1G OR30570-100MG OR30570-250MG
APOSOR30570-1GEA 11910 CZK
APOSOR30570-1G APOSOR30570-100MG APOSOR30570-250MG
6-(Bromomethyl)isoquinoline hydrobromide
6-(Bromomethyl)isoquinoline hydrobromide
Vzorec: C₁₀H₈BrN·BrH
Storage Temperature: Ambient
MDL Number: MFCD12022337
CAS číslo: 188861-57-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR