Tisk…

1H-Indol-4-ylacetic acid

Dodavatel: Apollo Scientific

Synonyms: 2-(1H-Indol-4-yl)acetic acid

OR305636-1G OR305636-250MG
APOSOR305636-1GEA 5390 CZK
APOSOR305636-1G APOSOR305636-250MG
1H-Indol-4-ylacetic acid
1H-Indol-4-ylacetic acid
Vzorec: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09837549
CAS číslo: 16176-74-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR