Tisk…

Methyl bromo(2-cyanophenyl)acetate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Methyl 2-bromo-2-(2-cyanophenyl)ethanoate, 2-(1-Bromo-2-methoxy-2-oxoethyl)benzonitrile

OR30563-100MG OR30563-1G OR30563-250MG
APOSOR30563-100MGEA 3970 CZK
APOSOR30563-100MG APOSOR30563-1G APOSOR30563-250MG
Methyl bromo(2-cyanophenyl)acetate
Methyl bromo(2-cyanophenyl)acetate
Vzorec: C₁₀H₈BrNO₂
MW: 254,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038664
CAS číslo: 885953-43-5
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR