Tisk…

1-Benzyl-D-proline 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (R)-1-Benzylpyrrolidine-2-carboxylic acid, N-D-Bzl-Pro-OH

OR305608-1G OR305608-5G
APOSOR305608-1GEA 1990 CZK
APOSOR305608-1G APOSOR305608-5G
1-Benzyl-D-proline 97%
1-Benzyl-D-proline
Vzorec: C₁₂H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD06656733
CAS číslo: 56080-99-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR