Tisk…

2,3-Dihydro-1H-pyrrolo[3,2-c]pyridine 95%

Dodavatel: Apollo Scientific
Varování

OR305601-1G OR305601-250MG
APOSOR305601-1GEA 13390 CZK
APOSOR305601-1G APOSOR305601-250MG
2,3-Dihydro-1H-pyrrolo[3,2-c]pyridine 95%
2,3-Dihydro-1H-pyrrolo[3,2-c]pyridine
Vzorec: C₇H₈N₂
MW: 120,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18382584
CAS číslo: 23596-28-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR